Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 117300977-117301138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROS1
Entrez Gene ID
6098
Gene Name
ROS proto-oncogene 1, receptor tyrosine kinase
Gene Aliases
MCF3, ROS, c-ros-1
UniGene
Hs.1041
Gene Chromosome Location
Chr.6: 117287918-117426065 on Build GRCh38
Transcripts
NM_002944.2 XM_006715548.3 XM_011536049.2 XM_011536050.2 XM_011536051.2 XM_011536052.2 XM_011536053.2 XM_011536054.2 XM_017011172.1 XM_017011173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4504401 c.6646C>T p.Q2216* Chr.6: 117301061-117301061 on Build GRCh38
3619811 c.6671G>A p.R2224K Chr.6: 117301018-117301018 on Build GRCh38
5404220 c.6655A>G p.S2219G Chr.6: 117301034-117301034 on Build GRCh38
7248054 c.6702T>G p.N2234K Chr.6: 117300987-117300987 on Build GRCh38
3157631 c.6621C>A p.D2207E Chr.6: 117301068-117301068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775088466 Chr.6: 117301114-117301114 on Build GRCh38
rs149949391 Chr.6: 117301088-117301088 on Build GRCh38
rs775605839 Chr.6: 117301047-117301047 on Build GRCh38
rs771313001 Chr.6: 117301097-117301097 on Build GRCh38
rs148104006 Chr.6: 117301078-117301078 on Build GRCh38
rs1051601581 Chr.6: 117301084-117301084 on Build GRCh38
rs773526509 Chr.6: 117301023-117301023 on Build GRCh38
rs145104466 Chr.6: 117301093-117301093 on Build GRCh38
rs376903822 Chr.6: 117301053-117301053 on Build GRCh38
rs748687432 Chr.6: 117301081-117301081 on Build GRCh38
rs529156 Chr.6: 117301025-117301025 on Build GRCh38
rs373160577 Chr.6: 117301059-117301059 on Build GRCh38
rs771437739 Chr.6: 117301004-117301005 on Build GRCh38
rs774048022 Chr.6: 117301033-117301033 on Build GRCh38
rs866580908 Chr.6: 117301073-117301073 on Build GRCh38
rs565633598 Chr.6: 117301027-117301027 on Build GRCh38
rs538165958 Chr.6: 117301080-117301080 on Build GRCh38
rs556427413 Chr.6: 117301100-117301100 on Build GRCh38
rs993857429 Chr.6: 117301058-117301058 on Build GRCh38
rs144594219 Chr.6: 117301096-117301096 on Build GRCh38
rs777946284 Chr.6: 117301036-117301036 on Build GRCh38
rs767110496 Chr.6: 117301043-117301043 on Build GRCh38
rs748632361 Chr.6: 117301009-117301009 on Build GRCh38
rs75510639 Chr.6: 117301068-117301068 on Build GRCh38
rs149121667 Chr.6: 117301095-117301095 on Build GRCh38
rs529038 Chr.6: 117301070-117301070 on Build GRCh38
rs745498493 Chr.6: 117301103-117301103 on Build GRCh38
rs1053611030 Chr.6: 117301007-117301007 on Build GRCh38
rs146680210 Chr.6: 117301108-117301108 on Build GRCh38
rs760646608 Chr.6: 117301048-117301048 on Build GRCh38
rs619203 Chr.6: 117301021-117301021 on Build GRCh38
rs759025806 Chr.6: 117301042-117301042 on Build GRCh38

View Full Product Details