Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 117310044-117310205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROS1
Entrez Gene ID
6098
Gene Name
ROS proto-oncogene 1, receptor tyrosine kinase
Gene Aliases
MCF3, ROS, c-ros-1
UniGene
Hs.1041
Gene Chromosome Location
Chr.6: 117287918-117426065 on Build GRCh38
Transcripts
NM_002944.2 XM_006715548.3 XM_011536049.2 XM_011536050.2 XM_011536051.2 XM_011536052.2 XM_011536053.2 XM_011536054.2 XM_017011172.1 XM_017011173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5576235 c.6311A>G p.Y2104C Chr.6: 117310186-117310186 on Build GRCh38
739517 c.6430G>A p.V2144I Chr.6: 117310085-117310085 on Build GRCh38
6972112 c.6416+1G>A p.? Chr.6: 117310080-117310080 on Build GRCh38
6836904 c.6352C>T p.P2118S Chr.6: 117310145-117310145 on Build GRCh38
1697814 c.6363G>A p.W2121* Chr.6: 117310134-117310134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749246490 Chr.6: 117310153-117310153 on Build GRCh38
rs754613413 Chr.6: 117310074-117310074 on Build GRCh38
rs886941681 Chr.6: 117310148-117310148 on Build GRCh38
rs559120791 Chr.6: 117310070-117310070 on Build GRCh38
rs749189360 Chr.6: 117310085-117310085 on Build GRCh38
rs150994413 Chr.6: 117310171-117310171 on Build GRCh38
rs748287694 Chr.6: 117310177-117310177 on Build GRCh38
rs138376257 Chr.6: 117310139-117310139 on Build GRCh38
rs141957047 Chr.6: 117310149-117310149 on Build GRCh38
rs745741484 Chr.6: 117310170-117310170 on Build GRCh38
rs770739791 Chr.6: 117310094-117310094 on Build GRCh38
rs752266755 Chr.6: 117310144-117310144 on Build GRCh38
rs762013317 Chr.6: 117310114-117310114 on Build GRCh38
rs199882276 Chr.6: 117310138-117310138 on Build GRCh38
rs757128233 Chr.6: 117310159-117310159 on Build GRCh38
rs376507701 Chr.6: 117310111-117310111 on Build GRCh38
rs954471241 Chr.6: 117310179-117310179 on Build GRCh38
rs761397175 Chr.6: 117310071-117310076 on Build GRCh38
rs778683218 Chr.6: 117310161-117310161 on Build GRCh38
rs373471832 Chr.6: 117310078-117310078 on Build GRCh38
rs773896715 Chr.6: 117310120-117310120 on Build GRCh38
rs774083623 Chr.6: 117310097-117310097 on Build GRCh38
rs747630349 Chr.6: 117310080-117310080 on Build GRCh38
rs765386753 Chr.6: 117310118-117310118 on Build GRCh38
rs777221228 Chr.6: 117310152-117310152 on Build GRCh38
rs747208320 Chr.6: 117310103-117310103 on Build GRCh38
rs145358991 Chr.6: 117310128-117310128 on Build GRCh38
rs1018655426 Chr.6: 117310137-117310137 on Build GRCh38
rs766995071 Chr.6: 117310165-117310166 on Build GRCh38
rs116038831 Chr.6: 117310083-117310083 on Build GRCh38
rs755564042 Chr.6: 117310081-117310081 on Build GRCh38
rs201206180 Chr.6: 117310174-117310174 on Build GRCh38

View Full Product Details