Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 78717271-78717544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL7
Entrez Gene ID
3574
Gene Name
interleukin 7
Gene Aliases
IL-7
UniGene
Hs.591873
Gene Chromosome Location
Chr.8: 78675870-78805523 on Build GRCh38
Transcripts
XM_011517523.2 NM_016010.2 XM_005251256.1

Gene Symbol
ZC2HC1A
Entrez Gene ID
51101
Gene Name
zinc finger C2HC-type containing 1A
Gene Aliases
C8orf70, CGI-62, FAM164A
UniGene
Hs.271876
Gene Chromosome Location
Chr.8: 78666047-78719765 on Build GRCh38
Transcripts
XM_011517523.2 NM_016010.2 XM_005251256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3925620 c.875C>T p.S292L Chr.8: 78717390-78717390 on Build GRCh38
5443570 c.933G>C p.W311C Chr.8: 78717448-78717448 on Build GRCh38
2876546 c.882A>C p.G294G Chr.8: 78717397-78717397 on Build GRCh38
1101878 c.901C>T p.H301Y Chr.8: 78717416-78717416 on Build GRCh38
3925621 c.900C>T p.C300C Chr.8: 78717415-78717415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781264276 Chr.8: 78717501-78717501 on Build GRCh38
rs767253867 Chr.8: 78717357-78717357 on Build GRCh38
rs767163652 Chr.8: 78717485-78717485 on Build GRCh38
rs1039383552 Chr.8: 78717323-78717323 on Build GRCh38
rs551023842 Chr.8: 78717503-78717503 on Build GRCh38
rs752591216 Chr.8: 78717512-78717513 on Build GRCh38
rs751109108 Chr.8: 78717353-78717353 on Build GRCh38
rs769574487 Chr.8: 78717466-78717466 on Build GRCh38
rs935845869 Chr.8: 78717364-78717364 on Build GRCh38
rs772869260 Chr.8: 78717468-78717468 on Build GRCh38
rs77219198 Chr.8: 78717351-78717351 on Build GRCh38
rs148584360 Chr.8: 78717477-78717477 on Build GRCh38
rs779146262 Chr.8: 78717371-78717371 on Build GRCh38
rs779352301 Chr.8: 78717501-78717502 on Build GRCh38
rs1054251539 Chr.8: 78717387-78717387 on Build GRCh38
rs375522431 Chr.8: 78717390-78717390 on Build GRCh38
rs750862330 Chr.8: 78717377-78717377 on Build GRCh38
rs780581410 Chr.8: 78717436-78717436 on Build GRCh38
rs747072248 Chr.8: 78717439-78717439 on Build GRCh38
rs754984633 Chr.8: 78717500-78717500 on Build GRCh38
rs368060254 Chr.8: 78717454-78717454 on Build GRCh38
rs750725772 Chr.8: 78717495-78717495 on Build GRCh38
rs775138408 Chr.8: 78717488-78717488 on Build GRCh38
rs760166877 Chr.8: 78717504-78717504 on Build GRCh38
rs760234371 Chr.8: 78717491-78717491 on Build GRCh38
rs769509602 Chr.8: 78717337-78717337 on Build GRCh38
rs201476525 Chr.8: 78717498-78717498 on Build GRCh38
rs749172086 Chr.8: 78717475-78717475 on Build GRCh38
rs752417374 Chr.8: 78717368-78717368 on Build GRCh38
rs748621985 Chr.8: 78717502-78717503 on Build GRCh38
rs778095421 Chr.8: 78717509-78717509 on Build GRCh38
rs776890062 Chr.8: 78717332-78717332 on Build GRCh38
rs770959996 Chr.8: 78717476-78717476 on Build GRCh38
rs5892663 Chr.8: 78717503-78717504 on Build GRCh38
rs757598027 Chr.8: 78717369-78717369 on Build GRCh38
rs758814241 Chr.8: 78717497-78717497 on Build GRCh38
rs769030515 Chr.8: 78717500-78717503 on Build GRCh38
rs141485055 Chr.8: 78717346-78717346 on Build GRCh38
rs763877690 Chr.8: 78717492-78717492 on Build GRCh38
rs749610268 Chr.8: 78717497-78717497 on Build GRCh38
rs747162119 Chr.8: 78717318-78717318 on Build GRCh38
rs761621155 Chr.8: 78717333-78717333 on Build GRCh38
rs748322481 Chr.8: 78717462-78717462 on Build GRCh38
rs759065173 Chr.8: 78717480-78717480 on Build GRCh38
rs759340186 Chr.8: 78717504-78717505 on Build GRCh38
rs781353383 Chr.8: 78717455-78717455 on Build GRCh38

View Full Product Details