Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2858702-2858947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXM1
Entrez Gene ID
2305
Gene Name
forkhead box M1
Gene Aliases
FKHL16, FOXM1B, HFH-11, HFH11, HNF-3, INS-1, MPHOSPH2, MPP-2, MPP2, PIG29, TRIDENT
UniGene
Hs.239
Gene Chromosome Location
Chr.12: 2857681-2877155 on Build GRCh38
Transcripts
NM_001243088.1 NM_001243089.1 NM_021953.3 NM_202002.2 NM_202003.2 XM_005253676.3 XM_011520930.2 XM_011520931.2 XM_011520934.2 NR_038933.1

Gene Symbol
LOC100507424
Entrez Gene ID
100507424
Gene Name
uncharacterized LOC100507424
Gene Aliases
-
UniGene
Hs.239
Gene Chromosome Location
Chr.12: 2836816-2859795 on Build GRCh38
Transcripts
NM_001243088.1 NM_001243089.1 NM_021953.3 NM_202002.2 NM_202003.2 XM_005253676.3 XM_011520930.2 XM_011520931.2 XM_011520934.2 NR_038933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5515463 c.2133G>A p.P711P Chr.12: 2858911-2858911 on Build GRCh38
212764 c.2313G>C p.K771N Chr.12: 2858731-2858731 on Build GRCh38
1322564 c.2136A>G p.Q712Q Chr.12: 2858908-2858908 on Build GRCh38
7332587 c.2169C>T p.I723I Chr.12: 2858875-2858875 on Build GRCh38
6666405 c.2260C>T p.R754C Chr.12: 2858784-2858784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138730141 Chr.12: 2858894-2858894 on Build GRCh38
rs140519202 Chr.12: 2858748-2858748 on Build GRCh38
rs200806960 Chr.12: 2858816-2858816 on Build GRCh38
rs28919869 Chr.12: 2858924-2858924 on Build GRCh38
rs150424596 Chr.12: 2858776-2858776 on Build GRCh38
rs753352536 Chr.12: 2858795-2858795 on Build GRCh38
rs749814710 Chr.12: 2858911-2858911 on Build GRCh38
rs757898539 Chr.12: 2858830-2858830 on Build GRCh38
rs150443540 Chr.12: 2858923-2858923 on Build GRCh38
rs775398816 Chr.12: 2858856-2858856 on Build GRCh38
rs759900571 Chr.12: 2858839-2858839 on Build GRCh38
rs767346257 Chr.12: 2858779-2858779 on Build GRCh38
rs771746117 Chr.12: 2858849-2858849 on Build GRCh38
rs548147648 Chr.12: 2858822-2858822 on Build GRCh38
rs369132152 Chr.12: 2858764-2858764 on Build GRCh38
rs748758384 Chr.12: 2858755-2858755 on Build GRCh38
rs369158837 Chr.12: 2858835-2858835 on Build GRCh38
rs758119183 Chr.12: 2858877-2858877 on Build GRCh38
rs778481065 Chr.12: 2858814-2858814 on Build GRCh38
rs975742881 Chr.12: 2858760-2858760 on Build GRCh38
rs769024368 Chr.12: 2858740-2858740 on Build GRCh38
rs964344748 Chr.12: 2858734-2858734 on Build GRCh38
rs765220332 Chr.12: 2858788-2858788 on Build GRCh38
rs778172822 Chr.12: 2858831-2858831 on Build GRCh38
rs376414243 Chr.12: 2858916-2858916 on Build GRCh38
rs769749986 Chr.12: 2858761-2858761 on Build GRCh38
rs749330783 Chr.12: 2858833-2858833 on Build GRCh38
rs748580011 Chr.12: 2858821-2858821 on Build GRCh38
rs751169233 Chr.12: 2858897-2858897 on Build GRCh38
rs749656919 Chr.12: 2858926-2858926 on Build GRCh38
rs759483249 Chr.12: 2858743-2858743 on Build GRCh38
rs146806643 Chr.12: 2858783-2858783 on Build GRCh38
rs781380281 Chr.12: 2858883-2858883 on Build GRCh38
rs756784859 Chr.12: 2858797-2858797 on Build GRCh38
rs28919870 Chr.12: 2858912-2858912 on Build GRCh38
rs753788611 Chr.12: 2858871-2858871 on Build GRCh38
rs189481330 Chr.12: 2858928-2858928 on Build GRCh38
rs995881166 Chr.12: 2858905-2858905 on Build GRCh38
rs747327902 Chr.12: 2858722-2858722 on Build GRCh38
rs778283602 Chr.12: 2858899-2858899 on Build GRCh38
rs144897055 Chr.12: 2858836-2858836 on Build GRCh38
rs138164446 Chr.12: 2858784-2858784 on Build GRCh38
rs763484794 Chr.12: 2858786-2858786 on Build GRCh38
rs903678903 Chr.12: 2858918-2858918 on Build GRCh38
rs773390706 Chr.12: 2858762-2858762 on Build GRCh38
rs113780841 Chr.12: 2858872-2858872 on Build GRCh38
rs201630079 Chr.12: 2858815-2858815 on Build GRCh38
rs563190488 Chr.12: 2858921-2858921 on Build GRCh38
rs745702333 Chr.12: 2858921-2858922 on Build GRCh38
rs770372738 Chr.12: 2858828-2858828 on Build GRCh38
rs760422215 Chr.12: 2858857-2858857 on Build GRCh38
rs764047099 Chr.12: 2858869-2858869 on Build GRCh38
rs764705130 Chr.12: 2858927-2858927 on Build GRCh38
rs753064494 Chr.12: 2858888-2858888 on Build GRCh38
rs763235392 Chr.12: 2858763-2858763 on Build GRCh38
rs941405297 Chr.12: 2858806-2858806 on Build GRCh38
rs774734794 Chr.12: 2858773-2858773 on Build GRCh38
rs764795562 Chr.12: 2858875-2858875 on Build GRCh38
rs145011914 Chr.12: 2858771-2858771 on Build GRCh38
rs781663757 Chr.12: 2858820-2858820 on Build GRCh38

View Full Product Details