Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008654.2 | NP_032680.1 | 3 | 2120 | |
GenBank mRNA | AK136336.1 | - | 3 | 2076 | |
AK154697.1 | - | 2 | 2602 | ||
AK167317.1 | - | 4 | 1845 | ||
AW493042.1 | - | 1 | 222 | ||
BC138485.1 | - | 3 | 1985 | 8861505 | |
BC138486.1 | - | 3 | 1985 | 8861506 | |
X51829.1 | - | 3 | 2065 |