Gene Transcripts

Gene Symbol
B3glct
Entrez Gene ID
381694
Gene Name
beta-3-glucosyltransferase
Gene Aliases
B3galtl, Gm1057
Chromosome Location
Chr. 5: 149678105 - 149762599 on Build GRCm38
UniGene ID
Mm.45060
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081204.1 NP_001074673.1 13 1141
GenBank mRNA AB253762.1 - 13 1106
AK145082.1 - 13 1116

View Full Product Details