Gene Transcripts

Gene Symbol
Ppp1r36
Entrez Gene ID
210762
Gene Name
protein phosphatase 1, regulatory subunit 36
Gene Aliases
Gm70
Chromosome Location
Chr. 12: 76417599 - 76439492 on Build GRCm38
UniGene ID
Mm.292354
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163103.1 NP_001156575.1 2 135
GenBank mRNA AK133227.1 - 2 114
CK137158.1 - 2 135

View Full Product Details