Gene Transcripts
- Gene Symbol
- Hist1h1d
- Entrez Gene ID
- 14957
- Gene Name
- histone cluster 1, H1d
- Gene Aliases
- H1.3, H1D, H1f3, H1s-4
- Chromosome Location
- Chr. 13: 23555032 - 23555807 on Build GRCm38
- UniGene ID
- Mm.247046
- Species
- Mus musculus
- Gene Symbol
- E430020m11rik
- Entrez Gene ID
- 100034673
- Gene Name
- RIKEN cDNA E430020M11 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.247046
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK088581.1 |
- |
1 |
513 |
|
View Full Product Details