Gene Transcripts

Gene Symbol
Hhat
Entrez Gene ID
226861
Gene Name
hedgehog acyltransferase
Gene Aliases
2810432O22Rik, AI462858, AP-2CRE, Skn, Tg(TFAP2A-cre)1Will
Chromosome Location
Chr. 1: 192512828 - 192771412 on Build GRCm38
UniGene ID
Mm.145857
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144881.4 NP_659130.2 6 803
GenBank mRNA AK028230.1 - 6 606
AK134661.1 - 6 647
AK172126.1 - 6 650
BC008159.1 - 797 3963716

View Full Product Details