Gene Transcripts

Gene Symbol
Meiob
Entrez Gene ID
75178
Gene Name
meiosis specific with OB domains
Gene Aliases
4930528F23Rik, MLZ-675
Chromosome Location
Chr. 17: 24804301 - 24839788 on Build GRCm38
UniGene ID
Mm.52592
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029197.1 NP_083473.1 11 1011
GenBank mRNA AK015924.1 - 3 252
BC119085.1 - 3 310 8733834
BC119087.1 - 3 310 8733836

View Full Product Details