Gene Transcripts

Gene Symbol
4930563E22Rik
Entrez Gene ID
75304
Gene Name
RIKEN cDNA 4930563E22 gene
Gene Aliases
Chromosome Location
Chr. 11: 72215138 - 72218450 on Build GRCm38
UniGene ID
Mm.340776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163728.1 NP_001157200.1 1 569
GenBank mRNA AK016202.1 - 1 379
AK043242.1 - 1 536
AK044443.1 - 1 570
AK132388.1 - 1 522

View Full Product Details