Gene Transcripts

Gene Symbol
RGD1561931
Entrez Gene ID
302396
Gene Name
similar to KIAA2022 protein
Gene Aliases
Chromosome Location
Chr. X: 74946745 - 75049602 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773350.1 XP_008771572.1 4 5573

View Full Product Details