Gene Transcripts

Gene Symbol
Micu2
Entrez Gene ID
171433
Gene Name
mitochondrial calcium uptake 2
Gene Aliases
Efha1, RGD1309934, Smhs2
Chromosome Location
Chr. 15: 38191506 - 38276159 on Build Rnor_6.0
UniGene ID
Rn.64644
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134396.2 NP_599223.2 10 985
GenBank mRNA DY317464.1 - 8 635

View Full Product Details