Gene Transcripts

Gene Symbol
Gpr157
Entrez Gene ID
313725
Gene Name
G protein-coupled receptor 157
Gene Aliases
Chromosome Location
Chr. 5: 167109691 - 167125380 on Build Rnor_6.0
UniGene ID
Rn.126639
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012107.1 NP_001012107.1 2 663
GenBank mRNA BC090013.1 - 2 663

View Full Product Details