Gene Transcripts

Gene Symbol
Bik
Entrez Gene ID
114496
Gene Name
BCL2-interacting killer (apoptosis-inducing)
Gene Aliases
Biklk, Blk
Chromosome Location
Chr. 7: 124390926 - 124410449 on Build Rnor_6.0
UniGene ID
Rn.38487
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053704.1 NP_446156.1 4 396
XM_006242048.2 XP_006242110.1 4 483
GenBank mRNA AF372501.1 - 4 396
FQ225338.1 - 4 373

View Full Product Details