Gene Transcripts

Gene Symbol
Trim28
Entrez Gene ID
116698
Gene Name
tripartite motif-containing 28
Gene Aliases
Kap1, Krip1, Tif1b
Chromosome Location
Chr. 1: 65544365 - 65551043 on Build Rnor_6.0
UniGene ID
Rn.198494
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053916.1 NP_446368.1 12 1719
XM_006228108.2 XP_006228170.1 12 1715
GenBank mRNA BC166843.1 - 12 1719

View Full Product Details