Gene Transcripts

Gene Symbol
RGD1307704
Entrez Gene ID
314300
Gene Name
similar to RIKEN cDNA 2410016O06
Gene Aliases
No66
Chromosome Location
Chr. 6: 107428508 - 107431126 on Build Rnor_6.0
UniGene ID
Rn.66167
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108040.1 NP_001101510.1 1 1266

View Full Product Details