Gene Transcripts

Gene Symbol
Ros1
Entrez Gene ID
19886
Gene Name
Ros1 proto-oncogene
Gene Aliases
Ros-1, c-ros
Chromosome Location
Chr.10: 52044865 - 52195397 on Build GRCm38
Uni Gene
Mm.236163
Interrogated Sequence Translated Protein
RefSeq NM_011282.2 NP_035412.2
GenBank mRNA U15443.1 -
X81650.1 -

View Full Product Details