Gene Transcripts
- Gene Symbol
- PRO0628
- Entrez Gene ID
- 29053
- Gene Name
- uncharacterized LOC29053
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.736997
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC015320.1 |
- |
1 |
762 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK093918.1 |
- |
1 |
26 |
|
Chr. 20 - 41038569 - 41040803 [+] |
View Full Product Details