Gene Transcripts
- Gene Symbol
- GAS2
- Entrez Gene ID
- 2620
- Gene Name
- growth arrest specific 2
- Gene Aliases
- Chromosome Location
- Chr. 11: 22626113 - 22885930 on Build GRCh38
- UniGene ID
- Hs.711297
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY027856.1 |
- |
1 |
231 |
|
Chr. 11 - 22625898 - 22626392 [+] |
GenBankmRNA |
BU561384.1 |
- |
1 |
190 |
|
Chr. 11 - 22625939 - 22626632 [+] |
GenBankmRNA |
BI253371.1 |
- |
1 |
186 |
|
Chr. 11 - 22625943 - 22626417 [+] |
View Full Product Details