Gene Transcripts
- Gene Symbol
- Gm11377
- Entrez Gene ID
- 105245413
- Gene Name
- predicted gene 11377
- Gene Aliases
- OTTMUSG00000000685
- Chromosome Location
- Chr. 13: 31564117 - 31567391 on Build GRCm38
- UniGene ID
- Mm.135634
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK085428.1 |
- |
|
529 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB500056.2 |
- |
4 |
533 |
|
Chr. 13 - 31564747 - 31573558 [+] |
GenBankmRNA |
BB661154.1 |
- |
4 |
526 |
|
Chr. 13 - 31564751 - 31573544 [+] |
View Full Product Details