Gene Transcripts
- Gene Symbol
- 2310021N16Rik
- Entrez Gene ID
- 72331
- Gene Name
- RIKEN cDNA 2310021N16 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK009452.1 |
- |
1 |
113 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY709178.1 |
- |
1 |
112 |
|
Chr. 2 - 148280475 - 148281157 [-] |
View Full Product Details