Gene Transcripts
- Gene Symbol
- 4930500F04Rik
- Entrez Gene ID
- 70969
- Gene Name
- RIKEN cDNA 4930500F04 gene
- Gene Aliases
- 4931407C17Rik
- Chromosome Location
- Chr. 5: 149847747 - 149864280 on Build GRCm38
- UniGene ID
- Mm.410799
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045758.1 |
|
1 |
188 |
|
GenBank mRNA |
AK015658.1 |
- |
1 |
188 |
|
View Full Product Details