Gene Transcripts
- Gene Symbol
- 1700022H16Rik
- Entrez Gene ID
- 69420
- Gene Name
- RIKEN cDNA 1700022H16 gene
- Gene Aliases
- Chromosome Location
- Chr. 12: 9565236 - 9570585 on Build GRCm38
- UniGene ID
- Mm.396193
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045488.1 |
|
4 |
345 |
|
GenBank mRNA |
AK006240.1 |
- |
4 |
346 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY706323.1 |
- |
4 |
346 |
|
Chr. 12 - 9565236 - 9570585 [-] |
View Full Product Details