Gene Transcripts
- Gene Symbol
- 4930563E22Rik
- Entrez Gene ID
- 75304
- Gene Name
- RIKEN cDNA 4930563E22 gene
- Gene Aliases
- Chromosome Location
- Chr. 11: 72215138 - 72218450 on Build GRCm38
- UniGene ID
- Mm.340776
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043242.1 |
- |
1 |
772 |
|
AK044443.1 |
- |
1 |
806 |
|
AK132388.1 |
- |
1 |
758 |
|
View Full Product Details