Gene Transcripts

Gene Symbol
Olfr1262
Entrez Gene ID
258976
Gene Name
olfactory receptor 1262
Gene Aliases
MOR234-1, or-leaf2
Chromosome Location
Chr. 2: 90002408 - 90003322 on Build GRCm38
UniGene ID
Mm.223367
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146974.1 NP_667185.1 1 519
GenBank mRNA AB074429.1 - 1 519
BC120560.1 - 1 580 40129483
BC120562.1 - 1 580 40129485

View Full Product Details