Gene Transcripts

Gene Symbol
Cinp
Entrez Gene ID
67236
Gene Name
cyclin-dependent kinase 2 interacting protein
Gene Aliases
1810047K05Rik, 2810452K22Rik
Chromosome Location
Chr. 12: 110872610 - 110889172 on Build GRCm38
UniGene ID
Mm.287157
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026048.4 NP_080324.1 4 432
NM_027223.1 NP_081499.1 5 582
GenBank mRNA AK004355.1 - 4 387
AK076126.1 - 4 402
AK160662.1 - 4 402
BC011066.1 - 4 402 3985202
BC021867.1 - 5 534 3710025

View Full Product Details