Gene Transcripts

Gene Symbol
Fgfr2
Entrez Gene ID
25022
Gene Name
fibroblast growth factor receptor 2
Gene Aliases
Chromosome Location
Chr. 1: 200590951 - 200696929 on Build Rnor_6.0
UniGene ID
Rn.12732
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109892.1 NP_001103362.1 13 2406
NM_001109893.1 NP_001103363.1 12 2142
NM_001109894.1 NP_001103364.1 12 2139
NM_001109895.1 NP_001103365.1 11 2064
NM_001109896.1 NP_001103366.1 11 2061
NM_012712.1 NP_036844.1 13 2409
XM_006230389.2 XP_006230451.1 13 2404
XM_006230390.2 XP_006230452.1 13 2401
XM_006230392.2 XP_006230454.1 13 2410
XM_006230393.2 XP_006230455.1 11 2056
XM_008759861.1 XP_008758083.1 12 4566
GenBank mRNA L19109.1 - 5 543
L19110.1 - 5 547
U59800.1 - 1 86
Z35138.1 - 10 1412
Z35139.1 - 9 1310

View Full Product Details