Gene Transcripts

Gene Symbol
Slc5a3
Entrez Gene ID
114507
Gene Name
solute carrier family 5 (sodium/myo-inositol cotransporter), member 3
Gene Aliases
Smit
Chromosome Location
Chr. 11: 32229366 - 32231812 on Build Rnor_6.0
UniGene ID
Rn.208396
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053715.2 NP_446167.2 1 1947
GenBank mRNA AY231162.1 - 1 1888

View Full Product Details