Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001173510.2 | NP_001166981.2 | 9 | 1183 | |
XM_006236193.2 | XP_006236255.1 | 10 | 1433 | ||
XM_006236194.2 | XP_006236256.1 | 9 | 1524 | ||
XM_008762770.1 | XP_008760992.1 | 9 | 1579 | ||
XM_008762771.1 | XP_008760993.1 | 9 | 1591 | ||
GenBank mRNA | CV113234.1 | - | 7 | 673 | |
FN804654.1 | - | 3 | 187 |