Gene Transcripts

Gene Symbol
Ercc6l2
Entrez Gene ID
361197
Gene Name
excision repair cross-complementation group 6-like 2
Gene Aliases
RGD1561537
Chromosome Location
Chr. 17: 1325653 - 1421732 on Build Rnor_6.0
UniGene ID
Rn.12539
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006253498.2 XP_006253560.2 2 523
GenBank mRNA FQ211532.1 - 2 470

View Full Product Details