Gene Transcripts

Gene Symbol
Avgr1
Entrez Gene ID
654840
Gene Name
autogenous vein graft remodeling associated protein 1
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.195307
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039588.1 NP_001034677.1 1703
GenBank mRNA DQ366675.1 - 1703

View Full Product Details