Gene Transcripts
- Gene Symbol
- Olfr665
- Entrez Gene ID
- 258810
- Gene Name
- olfactory receptor 665
- Gene Aliases
- MOR34-7
- Chromosome Location
- Chr. 7: 104880709 - 104881659 on Build GRCm38
- UniGene ID
- Mm.377700
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146814.1 |
NP_667025.1
|
1 |
751 |
|
View Full Product Details