Gene Transcripts
- Gene Symbol
- CUL1
- Entrez Gene ID
- 8454
- Gene Name
- cullin 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 148698178 - 148801110 on Build GRCh38
- UniGene ID
- Hs.146806
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003592.2 |
NP_003583.2
|
10, 11 |
1549 |
|
XM_005250060.4 |
XP_005250117.1
|
10, 11 |
1585 |
|
XM_011516629.2 |
XP_011514931.1
|
10, 11 |
1541 |
|
XM_011516630.2 |
XP_011514932.1
|
10, 11 |
1442 |
|
XM_011516631.2 |
XP_011514933.1
|
10, 11 |
1570 |
|
XM_011516632.2 |
XP_011514934.1
|
10, 11 |
1468 |
|
XM_017012723.1 |
XP_016868212.1
|
9, 10 |
1305 |
|
GenBank mRNA |
AF062536.1 |
- |
9, 10 |
1323 |
|
AK096163.1 |
- |
11, 12 |
1437 |
|
AK096505.1 |
- |
3, 4 |
1208 |
|
AK296909.1 |
- |
9, 10 |
1274 |
|
AK315023.1 |
- |
10, 11 |
1725 |
|
BC125119.1 |
- |
10, 11 |
1462 |
40118923 |
BC125120.1 |
- |
10, 11 |
1462 |
40118927 |
BX537409.1 |
- |
10, 11 |
1479 |
|
U58087.1 |
- |
10, 11 |
1251 |
|
View Full Product Details