Gene Transcripts

Gene Symbol
Vps13a
Entrez Gene ID
309243
Gene Name
vacuolar protein sorting 13 homolog A (S. cerevisiae)
Gene Aliases
RGD1311340
Chromosome Location
Chr. 1: 235352513 - 235813597 on Build Rnor_6.0
UniGene ID
Rn.16931
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100975.2 NP_001094445.2 54 7655
GenBank mRNA AY325251.1 - 11 2801

View Full Product Details