Gene Transcripts

Gene Symbol
Micu2
Entrez Gene ID
171433
Gene Name
mitochondrial calcium uptake 2
Gene Aliases
Efha1, RGD1309934, Smhs2
Chromosome Location
Chr. 15: 38191506 - 38276159 on Build Rnor_6.0
UniGene ID
Rn.64644
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134396.2 NP_599223.2 9 788
GenBank mRNA AF327513.1 - 9 788
DY317464.1 - 7 438
EV765275.1 - 9 796

View Full Product Details