Gene Transcripts

Gene Symbol
THOC7
Entrez Gene ID
80145
Gene Name
THO complex 7
Gene Aliases
NIF3L1BP1, fSAP24, hTREX30
Chromosome Location
Chr. 3: 63833870 - 63864478 on Build GRCh38
UniGene ID
Hs.288151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285387.2 NP_001272316.1 3 348
NM_001285404.1 NP_001272333.1 4 597
NM_025075.3 NP_079351.2 4 466
XM_006713339.3 XP_006713402.1 4 415
GenBank mRNA AF104914.1 - 3 314
AI391685.1 - 4 446
AI784549.1 - 3 346
AK027098.1 - 4 376
AK225110.1 - 4 354
BC020599.2 - 3 314
BC065012.1 - 4 336 6167514
BI914906.1 - 4 597
CR457373.1 - 4 333

View Full Product Details