Gene Transcripts
- Gene Symbol
- MAEL
- Entrez Gene ID
- 84944
- Gene Name
- maelstrom spermatogenic transposon silencer
- Gene Aliases
- CT128, SPATA35
- Chromosome Location
- Chr. 1: 166921121 - 167022214 on Build GRCh38
- UniGene ID
- Hs.651245
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286377.1 |
NP_001273306.1
|
4, 5 |
695 |
|
NM_001286378.1 |
NP_001273307.1
|
6, 7 |
561 |
|
NM_032858.2 |
NP_116247.1
|
5, 6 |
788 |
|
XM_006711583.1 |
XP_006711646.1
|
6, 7 |
592 |
|
XM_011510067.1 |
XP_011508369.1
|
5, 6 |
754 |
|
XM_011510068.1 |
XP_011508370.1
|
6, 7 |
545 |
|
XM_017002602.1 |
XP_016858091.1
|
7, 8 |
929 |
|
XM_017002603.1 |
XP_016858092.1
|
6, 7 |
591 |
|
GenBank mRNA |
AK027810.1 |
- |
5, 6 |
595 |
|
AK131066.1 |
- |
5, 6 |
598 |
|
AK302253.1 |
- |
4, 5 |
511 |
|
AL133073.1 |
- |
4, 5 |
352 |
|
BC028595.2 |
- |
5, 6 |
568 |
5270252 |
BC034310.1 |
- |
4, 5 |
1073 |
|
DQ076156.2 |
- |
5, 6 |
597 |
|
GU383113.1 |
- |
4, 5 |
504 |
|
GU383114.1 |
- |
6, 7 |
561 |
|
View Full Product Details