Gene Transcripts

Gene Symbol
OR8B12
Entrez Gene ID
219858
Gene Name
olfactory receptor family 8 subfamily B member 12
Gene Aliases
OR11-317
Chromosome Location
Chr. 11: 124542722 - 124543654 on Build GRCh38
UniGene ID
Hs.553634
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005195.1 NP_001005195.1 1 768
GenBank mRNA BC136863.1 - 1 793
BC136864.1 - 1 793 9020855

View Full Product Details