Gene Transcripts

Gene Symbol
KCNJ10
Entrez Gene ID
3766
Gene Name
potassium voltage-gated channel subfamily J member 10
Gene Aliases
BIRK-10, KCNJ13-PEN, KIR1.2, KIR4.1, SESAME
Chromosome Location
Chr. 1: 160037467 - 160070261 on Build GRCh38
UniGene ID
Hs.408960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002241.4 NP_002232.2 2 355
GenBank mRNA AK312383.1 - 2 262
BC034036.1 - 2 257
BC131627.1 - 2 251 40118533
DA958759.1 - 2 355
U52155.1 - 2 341

View Full Product Details