Gene Transcripts
- Gene Symbol
- EXOSC3
- Entrez Gene ID
- 51010
- Gene Name
- exosome component 3
- Gene Aliases
- CGI-102, PCH1B, RRP40, Rrp40p, bA3J10.7, hRrp-40, p10
- Chromosome Location
- Chr. 9: 37779714 - 37785092 on Build GRCh38
- UniGene ID
- Hs.602571
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001002269.2 |
NP_001002269.1
|
1, 2 |
380 |
|
NM_016042.3 |
NP_057126.2
|
1, 2 |
380 |
|
GenBank mRNA |
AF151860.1 |
- |
1, 2 |
306 |
|
AF229833.1 |
- |
1, 2 |
77 |
|
AF281132.1 |
- |
1, 2 |
356 |
|
AK289571.1 |
- |
1, 2 |
349 |
|
AK290864.1 |
- |
1, 2 |
345 |
|
BC002437.2 |
- |
1, 2 |
344 |
3346075 |
BC008880.2 |
- |
1, 2 |
334 |
4300826 |
BP237222.1 |
- |
1, 2 |
379 |
|
CF994051.1 |
- |
1, 2 |
388 |
|
DA352003.1 |
- |
1, 2 |
354 |
|
View Full Product Details