Gene Transcripts
- Gene Symbol
- LOC105372038
- Entrez Gene ID
- 105372038
- Gene Name
- uncharacterized LOC105372038
- Gene Aliases
- Chromosome Location
- Chr. 18: 27336991 - 27342656 on Build GRCh38
- UniGene ID
- Hs.722107
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_134589.1 |
|
2 |
191 |
|
GenBank mRNA |
DB447025.1 |
- |
2 |
193 |
|
View Full Product Details