Gene Transcripts
- Gene Symbol
- IGBP1
- Entrez Gene ID
- 3476
- Gene Name
- immunoglobulin (CD79A) binding protein 1
- Gene Aliases
- ALPHA-4, IBP1
- Chromosome Location
- Chr. X: 70133449 - 70166324 on Build GRCh38
- UniGene ID
- Hs.496267
- Species
- Homo sapiens
- Gene Symbol
- IGBP1P1
- Entrez Gene ID
- 280655
- Gene Name
- immunoglobulin (CD79A) binding protein 1 pseudogene 1
- Gene Aliases
- C14orf19
- Chromosome Location
- Chr. 14: 34939922 - 34940496 on Build GRCh38
- UniGene ID
- Hs.639271
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002937.2 |
|
1 |
127 |
|
GenBank mRNA |
AY168620.1 |
- |
1 |
127 |
|
View Full Product Details