Gene Transcripts

Gene Symbol
Mamdc2
Entrez Gene ID
71738
Gene Name
MAM domain containing 2
Gene Aliases
1200015L10Rik, AI591564, mamcan
Chromosome Location
Chr. 19: 23302609 - 23448500 on Build GRCm38
UniGene ID
Mm.50841
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174857.3 NP_777282.1 9 1553
GenBank mRNA AK004794.1 - 8 1416
AK035566.1 - 9 1554
AK154219.1 - 9 1630

View Full Product Details