Gene Transcripts

Gene Symbol
HOXC6
Entrez Gene ID
3223
Gene Name
homeobox C6
Gene Aliases
CP25, HHO.C8, HOX3, HOX3C
Chromosome Location
Chr. 12: 54016852 - 54030823 on Build GRCh38
UniGene ID
Hs.549040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004503.3 NP_004494.1 1 413
NM_153693.4 NP_710160.1 2 810
GenBank mRNA AK314829.1 - 2 490
BC074844.2 - 1 327 30915460
BC074845.2 - 1 327 30915223
CR456954.1 - 1 301
M16938.1 - 2 537
S82986.1 - 1 433

View Full Product Details