Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037808.2 | NP_001032897.1 | 9 | 1021 | |
NM_001199142.1 | NP_001186071.1 | 9 | 1141 | ||
NM_001267574.2 | NP_001254503.1 | 8 | 1110 | ||
NM_001286478.1 | NP_001273407.1 | 9 | 942 | ||
NM_003752.4 | NP_003743.1 | 9 | 972 | ||
XM_017023814.1 | XP_016879303.1 | 9 | 927 | ||
GenBank mRNA | AK000739.1 | - | 8 | 1002 | |
AK055366.1 | - | 9 | 841 | ||
AK130531.1 | - | 2 | 169 | ||
AK292155.1 | - | 9 | 1141 | ||
AK293264.1 | - | 7 | 550 | ||
AK299421.1 | - | 4, 5 | 381 | ||
AK301183.1 | - | 8 | 880 | ||
AK303790.1 | - | 9 | 885 | ||
AK304500.1 | - | 8 | 651 | ||
AY762099.1 | - | 9 | 884 | ||
BC001571.1 | - | 9 | 906 | 3452953 | |
BC071705.1 | - | 9 | 900 | 5552014 | |
BC157842.1 | - | 9 | 1030 | 9057061 | |
BC157849.1 | - | 8 | 870 | 9057068 | |
BX648075.1 | - | 1 | 1973 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099661.2 | NP_001093131.1 | 9 | 973 | |
NM_001317856.1 | NP_001304785.1 | 9 | 1018 | ||
NM_001317857.1 | NP_001304786.1 | 9 | 969 | ||
XM_017023620.1 | XP_016879109.1 | 9 | 925 | ||
XM_017023621.1 | XP_016879110.1 | 2 | 100 |