Gene Transcripts

Gene Symbol
Zfp459
Entrez Gene ID
328274
Gene Name
zinc finger protein 459
Gene Aliases
9930025G17Rik, Rslcan-14
Chromosome Location
Chr. 13: 67405712 - 67443302 on Build GRCm38
UniGene ID
Mm.222470
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177811.4 NP_808479.1 4 955
GenBank mRNA AK036923.1 - 4 956
BC100389.1 - 4 922

View Full Product Details