Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077509.1 | NP_001070977.1 | 7, 8 | 789 | |
NM_011612.2 | NP_035742.1 | 7, 8 | 808 | ||
GenBank mRNA | AK019885.1 | - | 7, 8 | 775 | |
AK154625.1 | - | 7, 8 | 792 | ||
DQ832278.1 | - | 6, 7 | 694 |