Gene Transcripts

Gene Symbol
Dlg2
Entrez Gene ID
23859
Gene Name
discs, large homolog 2 (Drosophila)
Gene Aliases
A330103J02Rik, B230218P12Rik, B330007M19Rik, Dlgh2, Gm1197, PSD93
Chromosome Location
Chr. 7: 90476183 - 92449246 on Build GRCm38
UniGene ID
Mm.147213
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011807.3 NP_035937.2 8 1064
GenBank mRNA AF388675.1 - 8 843
AK046525.1 - 7 1104

View Full Product Details