Gene Transcripts

Gene Symbol
Myo9a
Entrez Gene ID
171296
Gene Name
myosin IXA
Gene Aliases
Chromosome Location
Chr. 8: 64573248 - 64777607 on Build Rnor_6.0
UniGene ID
Rn.88901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134335.1 NP_599162.1 2 549
GenBank mRNA AJ001713.1 - 2 549

View Full Product Details